Learn More
USA Scientific Inc Tough-Spots, 1.0 inch dia
Catalog No. 502744963
Encompass
Tough-Spots, white labels for 50 mL tubes, 1.0 inch diameter, 500/roll. For -196C to +80C.
Catalog No. 50-274-4963 Supplier USA Scientific Inc Supplier No. 91850100
Tough-Spots, white labels for 50 mL tubes, 1.0 inch diameter, 500/roll. For -196C to +80C.