Learn More
USA Scientific Inc Tough-Spots, 1/2" dia, rolls
Catalog No. 502745530
Encompass
Tough-Spots, large, maroon labels for 1.5 mL and 2.0 mL tubes, 1/2 inch diameter, 1000/roll. For -196C to +80C.
Catalog No. 50-274-5530 Supplier USA Scientific Inc Supplier No. 91850514
Tough-Spots, large, maroon labels for 1.5 mL and 2.0 mL tubes, 1/2 inch diameter, 1000/roll. For -196C to +80C.