Learn More
USA Scientific Inc Tough-Spots, 1/2" dia, silver
Catalog No. 502745464
Encompass
Tough-Spots, large, silver labels for 1.5 mL and 2.0 mL tubes, 1/2 inch diameter, 1000/roll. For -196C to +80C.
Catalog No. 50-274-5464 Supplier USA Scientific Inc Supplier No. 91850518
Tough-Spots, large, silver labels for 1.5 mL and 2.0 mL tubes, 1/2 inch diameter, 1000/roll. For -196C to +80C.