Learn More
USA Scientific Inc Tough-Spots, 1/2 in dia
Catalog No. 502745511
Encompass
Tough-Spots, large, white labels for 1.5 mL and 2.0 mL tubes, 1/2 inch diameter, 1000/roll. For -196C to +80C.
Catalog No. 50-274-5511 Supplier USA Scientific Inc Supplier No. 91850500
Tough-Spots, large, white labels for 1.5 mL and 2.0 mL tubes, 1/2 inch diameter, 1000/roll. For -196C to +80C.