Learn More
Research Products International Corp Tough-Spots Multi-Roll, 3/8"" Diameter, Assorted Colors, 5 Labels Across, 5,000 Labels per Roll
SDP
Supplier:  Research Products International Corp 247160
Encompass
Convenient five across labels available in assorted colors on same roll for quick and easy color coding of sample tubes. Supplied in a cube style dispenser box that keeps labels neat and clean and ready for use. Tough-Spots withstand temperature down to -196°C without cracking or peeling and fit all 0.5-2.0ml micro-tubes. Labels are resistant to organic solvents and caustic agents.
Catalog No. 50-550-261