Learn More
Research Products International Corp Tough-Spots, 3/8 Inch Diameter for 0.5-2.0ml Tubes, Blue, 1,000 per Package
SDP
Catalog No. 50550366
Encompass
Tough-Spots are chemically resistant labels that adhere to polypropylene tubes without cracking, peeling or degrading. Specially designed to withstand boiling water baths, autoclaving freezing, cryogenic liquid nitrogen storage, organic solvents and caustic agents. Tough-Spots are made of a special smear resistant material and will accept any marking instrument.
Catalog No. 50-550-366 Supplier Research Products International Corp Supplier No. 247106B
Tough-Spots are chemically resistant labels that adhere to polypropylene tubes without cracking, peeling or degrading. Specially designed to withstand boiling water baths, autoclaving freezing, cryogenic liquid nitrogen storage, organic solvents and caustic agents. Tough-Spots are made of a special smear resistant material and will accept any marking instrument.