Learn More
Andwin Scientific TOUGHSPOTS ASSTD 1/2 5K/PK
SDP
Catalog No. 50727723
Encompass_Preferred
Suitable for 1.5-2ml microcentrifuge tube tops with 1/2 in. diameter, the peel-off labels are chemical and temperature resistant from -196-150 deg.C. It comes in a sheet of 5000 in 5 assorted colors.
Catalog No. 50-727-723 Supplier Andwin Scientific Supplier No. 7758621
Suitable for 1.5-2ml microcentrifuge tube tops with 1/2 in. diameter, the peel-off labels are chemical and temperature resistant from -196-150 deg.C. It comes in a sheet of 5000 in 5 assorted colors.