Learn More
Andwin Scientific TOUGHSPOTS ASSTD 3/8 5K/PK
SDP
Catalog No. 50727721
Encompass_Preferred
Suitable for 0.5ml microcentrifuge tube tops with 3/8 in. diameter, the peel-off labels are chemical and temperature resistant from -196-150 deg.C. It comes in a sheet of 5000 in 5 assorted colors.
Catalog No. 50-727-721 Supplier Andwin Scientific Supplier No. 7758611
Suitable for 0.5ml microcentrifuge tube tops with 3/8 in. diameter, the peel-off labels are chemical and temperature resistant from -196-150 deg.C. It comes in a sheet of 5000 in 5 assorted colors.