Learn More
USA Scientific Inc Tough-Spots, LG, gold
Catalog No. 502745456
Encompass
Tough-Spots, large, gold labels for 1.5 mL and 2.0 mL tubes, 1/2 inch diameter, 1000/roll. For -196C to +80C.
Catalog No. 50-274-5456 Supplier USA Scientific Inc Supplier No. 91850516
Tough-Spots, large, gold labels for 1.5 mL and 2.0 mL tubes, 1/2 inch diameter, 1000/roll. For -196C to +80C.