Learn More
Research Products International Corp Tough-Spots Multi-Roll, 1/2"" Diameter, Assorted Colors, 5 Labels Across, 5,000 Labels per Roll
SDP
Catalog No. 50550263
Encompass
Convenient five across labels available in assorted colors on same roll for quick and easy color coding of sample tubes. Supplied in a cube style dispenser box that keeps labels neat and clean and ready for use. Tough-Spots withstand temperature down to -196°C without cracking or peeling and fit all 0.5-2.0ml micro-tubes. Labels are resistant to organic solvents and caustic agents.
Catalog No. 50-550-263 Supplier Research Products International Corp Supplier No. 247162
Convenient five across labels available in assorted colors on same roll for quick and easy color coding of sample tubes. Supplied in a cube style dispenser box that keeps labels neat and clean and ready for use. Tough-Spots withstand temperature down to -196°C without cracking or peeling and fit all 0.5-2.0ml micro-tubes. Labels are resistant to organic solvents and caustic agents.