Learn More
Andwin Scientific TOUGHSPOTS RL 1/2 GR 1K/PK
SDP
Catalog No. 50727708
Encompass_Preferred
The Tough-Spots labels on a roll are specially designed to be used for 2.0 mL tubes. The labels are resistant against chemicals and temperatures from -196C to 80C. This product can be put on several surfaces like most plastics, glass, vinyl, and metals. Any marking instrument can also be utilized to write on these labels. This product has a diameter of 1/2". It is sold in a pack of 1000 green labels.
Catalog No. 50-727-708 Supplier Andwin Scientific Supplier No. 77581G
The Tough-Spots labels on a roll are specially designed to be used for 2.0 mL tubes. The labels are resistant against chemicals and temperatures from -196C to 80C. This product can be put on several surfaces like most plastics, glass, vinyl, and metals. Any marking instrument can also be utilized to write on these labels. This product has a diameter of 1/2". It is sold in a pack of 1000 green labels.