Learn More
Andwin Scientific TOUGHSPOTS RL 1/2 OR 1K/PK
SDP
Catalog No. 50727710
Encompass_Preferred
Suitable for 1.5-2ml microcentrifuge tube tops with 1/2 in. diameter, the pre-cut peel-off round PVC labels are easy to write on and is compatible with most lab markers. It comes in a roll of 1000 in color Orange.
Catalog No. 50-727-710 Supplier Andwin Scientific Supplier No. 77581O
Suitable for 1.5-2ml microcentrifuge tube tops with 1/2 in. diameter, the pre-cut peel-off round PVC labels are easy to write on and is compatible with most lab markers. It comes in a roll of 1000 in color Orange.