Learn More
Andwin Scientific TOUGHSPOTS RL 3/8 YL 1K/PK
SDP
Catalog No. 50727706
Encompass_Preferred
Suitable for 0.5ml microcentrifuge tube tops with 3/8 in. diameter, the pre-cut peel-off round PVC labels are easy to write on and is compatible with most lab markers. It comes in a roll of 1000 in color Yellow.
Catalog No. 50-727-706 Supplier Andwin Scientific Supplier No. 77580Y
Suitable for 0.5ml microcentrifuge tube tops with 3/8 in. diameter, the pre-cut peel-off round PVC labels are easy to write on and is compatible with most lab markers. It comes in a roll of 1000 in color Yellow.