Learn More
Andwin Scientific TOUGHSPOTS SH 1/2 RBW 3840/PK
SDP
Catalog No. 50727724
Encompass_Preferred
The Tough-Spots labels on a roll are specially designed to be used for 2.0 mL tubes. The labels are resistant against chemicals and temperatures from -196C to 80C. This product can be put on several surfaces like most plastics, glass, vinyl, and metals. Any marking instrument can also be utilized to write on these labels. This product has a diameter of 1/2". It is sold in a pack of 3840 labels in the color of blue, green, orange, red, and yellow.
Catalog No. 50-727-724 Supplier Andwin Scientific Supplier No. 7758705
The Tough-Spots labels on a roll are specially designed to be used for 2.0 mL tubes. The labels are resistant against chemicals and temperatures from -196C to 80C. This product can be put on several surfaces like most plastics, glass, vinyl, and metals. Any marking instrument can also be utilized to write on these labels. This product has a diameter of 1/2". It is sold in a pack of 3840 labels in the color of blue, green, orange, red, and yellow.