Learn More
Andwin Scientific TOUGHSPOTS SH 1/2 WH 3840/PK
SDP
Catalog No. 50727727
Encompass_Preferred
Suitable for 1.5-2ml microcentrifuge tube tops with 1/2 in. diameter, the peel-off labels are chemical and temperature resistant from -196-150 deg.C. It is made with polyolefin and acrylic adhesive and comes in a sheet of 3840 in color White.
Catalog No. 50-727-727 Supplier Andwin Scientific Supplier No. 77587W
Suitable for 1.5-2ml microcentrifuge tube tops with 1/2 in. diameter, the peel-off labels are chemical and temperature resistant from -196-150 deg.C. It is made with polyolefin and acrylic adhesive and comes in a sheet of 3840 in color White.