Learn More
Andwin Scientific TOUGHSPOTS SH 3/8 BL 3840/PK
SDP
Catalog No. 50727715
Encompass_Preferred
Suitable for 0.5ml microcentrifuge tube tops with 3/8 in. diameter, the peel-off labels are chemical and temperature resistant from -196-150 deg.C. It is made with polyolefin and acrylic adhesive and comes in a sheet of 3840 in color Blue.
Catalog No. 50-727-715 Supplier Andwin Scientific Supplier No. 77585B
Suitable for 0.5ml microcentrifuge tube tops with 3/8 in. diameter, the peel-off labels are chemical and temperature resistant from -196-150 deg.C. It is made with polyolefin and acrylic adhesive and comes in a sheet of 3840 in color Blue.