Learn More
Andwin Scientific TOUGHSPOTS SH 7/16 WH 4680/PK
SDP
Catalog No. 50727714
Encompass_Preferred
Suitable for 1.5-2ml microcentrifuge tube tops with 7/16 in. diameter, the peel-off labels are chemical and temperature resistant from -196-150 deg.C. It is made with polyolefin and acrylic adhesive and comes in a sheet of 4680 in color White.
Catalog No. 50-727-714 Supplier Andwin Scientific Supplier No. 7758544
Suitable for 1.5-2ml microcentrifuge tube tops with 7/16 in. diameter, the peel-off labels are chemical and temperature resistant from -196-150 deg.C. It is made with polyolefin and acrylic adhesive and comes in a sheet of 4680 in color White.