Learn More
Andwin Scientific TOUGHSPOTS WH 1/2 5K/PK
SDP
Catalog No. 50727722
Encompass_Preferred
Suitable for 1.5-2ml microcentrifuge tube tops with 1/2 in. diameter, the peel-off labels are chemical and temperature resistant from -196-150 deg.C. It comes in a sheet of 5000 in White.
Catalog No. 50-727-722 Supplier Andwin Scientific Supplier No. 7758620
Suitable for 1.5-2ml microcentrifuge tube tops with 1/2 in. diameter, the peel-off labels are chemical and temperature resistant from -196-150 deg.C. It comes in a sheet of 5000 in White.