Learn More
Brady™ ToughStripe Floor Tape Applicator
Catalog No. 17985484 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black, White, Yellow
Catalog No. Color
17-985-484 Black, White, Yellow
1 options
Catalog No. 17-985-484 Supplier Brady™ Supplier No. 178687
Add to Cart
Add to Cart
Comfortably lay straighter lines in less time with an ergonomic design, removable cartridge, and easy-apply handling features.
  • Patented removable cartridge: Simply pop it out, load tape, and snap it back in to effortlessly load tape at a comfortable height.
  • Optimized for single-person use: The easy-to-use, lightweight design allows one person to complete the work of two or three, improving productivity and efficiency.
  • Stable three-wheel design: Provides consistently straight, professional-grade lines with superior control, balance, and stability.
  • Patented tightening disc: Ensures secure tape handling and consistent payout, enabling smooth, straight, and jam-free application from start to finish.
  • Integrated liner rewinder: Features a patented rubber grip that automatically collects spent tape liner, keeping the workspace neat and organized.
  • Removable push handle and carrying handle: Push handle detaches for convenient storage, while the carrying handle enables easy lifting and transport of the applicator.
  • Built-in utility compartment: Provides storage for essential tools such as scissors and utility knives, keeping them within reach during operation.
  • Versatile tape compatibility: Works efficiently with any tape ranging from 2 to 6 in. in width, making it suitable for a variety of floor marking needs.
Specifications
Material Plastic, Steel
Product Type Floor Tape Applicator
Color Black, White, Yellow