Learn More
MSC TOUGHSTRIPE FLR MARK TAPE
Catalog No. 5000769908
Encompass
ToughStripe® Floor Tape provides effective identification, easy application, and uncompromising durability for maintaining a safe and efficient workplace. These tapes are designed to adhere to various challenging floor conditions, ensuring compliance with OSHA and ANSI standards for floor marking.
  • Ideal for heavy foot traffic, light industrial, and Lean/5S applications
  • Constructed from durable polyester material
  • Withstands temperatures from 50°F to 140°F
  • Features a thickness of 0.008 in.
  • Minimum application temperature of 50°F
Catalog No. 50-007-69908 Supplier MSC Supplier No. 10909141
ToughStripe® Floor Tape provides effective identification, easy application, and uncompromising durability for maintaining a safe and efficient workplace. These tapes are designed to adhere to various challenging floor conditions, ensuring compliance with OSHA and ANSI standards for floor marking.
  • Ideal for heavy foot traffic, light industrial, and Lean/5S applications
  • Constructed from durable polyester material
  • Withstands temperatures from 50°F to 140°F
  • Features a thickness of 0.008 in.
  • Minimum application temperature of 50°F