Learn More
MSC 3M Petroleum Sorbent Mini-Boom, Polypropylene, 3 in. x 8 ft., 6/Pk.
Catalog No. 5000687511
Encompass
These petroleum sorbent mini-booms are designed to control hydrocarbon and non-polar organic solvent spills on land and water. Constructed from inert synthetic fibers, including polypropylene, these sorbents repel water and float, making them suitable for use in ditches, streams, rivers, and docks. They feature a unique double-cylinder design to maximize absorption while minimizing oil escape in flowing water.
  • Provides high absorption capacity to minimize waste disposal
  • Features UV-resistant materials that do not deteriorate or disintegrate
  • Includes built-in attachment clips at each end to allow linking multiple booms together
  • Secured by netting to contain particulate sorbent material
  • Intended for use in low-temperature and low-humidity environments
  • Suitable for placing around equipment to prevent leaks and spills from spreading
Catalog No. 50-006-87511 Supplier MSC Supplier No. 59055897
These petroleum sorbent mini-booms are designed to control hydrocarbon and non-polar organic solvent spills on land and water. Constructed from inert synthetic fibers, including polypropylene, these sorbents repel water and float, making them suitable for use in ditches, streams, rivers, and docks. They feature a unique double-cylinder design to maximize absorption while minimizing oil escape in flowing water.
  • Provides high absorption capacity to minimize waste disposal
  • Features UV-resistant materials that do not deteriorate or disintegrate
  • Includes built-in attachment clips at each end to allow linking multiple booms together
  • Secured by netting to contain particulate sorbent material
  • Intended for use in low-temperature and low-humidity environments
  • Suitable for placing around equipment to prevent leaks and spills from spreading