Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Tough-Tag Labels On a Roll, 24 mm x 13 mm, Yellow
SDP

Supplier:  Electron Microscopy Sciences 62044Y

Encompass_Preferred

Tough-Tags are pre-cut, peel-off labels that accept any marking instrument. They are perfectly sized to fit microcentrifuge tubes and other containers in the laboratory. Dispensing box assists in labeling individual tubes used in small experiments.
Size: 24 mm x 13 mm
Color: Yellow
Use: 0.5 mL Tubes

Catalog No. 50-193-0979


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.