Learn More
Research Products International Corp Tough-Tags, 0.94 x 0.5 Inch Labels for 0.5-0.65ml Tubes, Clear, 1000 per Package
SDP
Supplier:  Research Products International Corp 247100
Encompass
Tough-Tags are pre-cut, peel-off labels that fit micro-tubes and a variety of other containers in research laboratories. Labels are manufactured from chemically inert polyester with strong adhesion properties. Tough-Tags adhere to all plastics and withstand steam autoclaving, boiling water baths, freezer temperatures, organic solvents and caustic agents.
Not recommended for use with liquid nitrogen.
Tough-Tags are made of a special smear resistant material and will accept any marking instrument.
Catalog No. 50-550-242