Learn More
Andwin Scientific 38X19mm Laser Tough-Tags Labels, White, 1500 pack
SDP
Supplier:  Andwin Scientific 77522W
Encompass_Preferred
For general purposes, the 38X19mm Laser Tough-Tags Labels can withstand temperatures ranging from -40-121 deg.C. These are stay-flat labels for easy laser printing that comes in color White in a pack of 1500.
Catalog No. 50-727-667