Learn More
Research Products International Corp Tough-Tags Sheets for Laser Printers, 1.5ml Tubes, 1.28 x 0.5 Inches, White, 2,520 per Package
SDP
Supplier:  Research Products International Corp 247105
Encompass
Tough Tags are pre-cut, peel-off labels that fit micro-tubes and a variety of other containers in research laboratories. Labels are manufactured from chemically inert polyester with strong adhesion properties. The labels adhere to all plastics and withstand steam autoclaving, boiling water baths, freezer temperatures, organic solvents, and caustic agents. Not recommended for use with liquid nitrogen. Tough-Tags are made of a special smear resistant material and will accept any marking instrument.
Catalog No. 50-550-246