Learn More
3M™ Tour-Guard™ V Protective Eyewear
Catalog No. 19039591 Shop All 3M Products
Change view
Click to view available options
No. per Pack:
20/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
19-039-591 20/Pk.
19-039-590 20/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-039-591 Supplier 3M™ Supplier No. 7100086111
  • PRODUCT PROMOTION
    $611.00
    $243.02 / Case of 100
    Save $367.98
    60% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
3M™ Tour-Guard™ V Protective Eyewear offers impact protection, integral sideshields and flexible, polycarbonate construction with over-the-glass coverage.
  • Economical protective eyewear serves needs of site visitors or general use
  • Wraparound side shields help provide extra protection
  • Medium size fits over most personal prescription eyewear
  • Polycarbonate lenses absorb 99.9% UVA and UVB up to 380 nm
  • 3M Catalog# TGV01-100 , 3M Catalog# TGV01-20
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating UV-treated
Certifications/Compliance ANSI Z87.1-2015
Lens Tint Clear
Product Type Safety Glasses
Frame Color Clear
Model TGV01-100
Quantity 100 Ea.
No. per Pack 20/Pk.
Temple Style Classic
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV400
Frame Type Frameless