Learn More
Med Vet International C-A-T Trainer Tourniquet 37.5 Inch Length Military Latex Free Nylon, 30-0033, Each
Catalog No. 503043437
Encompass_Preferred
Single Routing Buckle for faster application, decreased blood loss, effective slack removal, fewer windlass turns, and simplified training with single protocol application standards Reinforced windlass rod with increased diameter for enhanced strength and aggressive ribbing for improved grip Bilateral windlass clip for rapid windlass lock and bilateral buttress for added strength Enhanced windlass strap changed to tactical gray and secured to the latch for constant contact providing safe windlass security and a place to document the application time Reinforced stabilization plate with beveled contact bar Signature red elliptical tip (Red Tip Technology) provides visual cue during application Providing a distinct visual means to distinguish training devices from personal/operational life-saving devices
Catalog No. 50-304-3437 Supplier Med Vet International Supplier No. 812822EA
Single Routing Buckle for faster application, decreased blood loss, effective slack removal, fewer windlass turns, and simplified training with single protocol application standards Reinforced windlass rod with increased diameter for enhanced strength and aggressive ribbing for improved grip Bilateral windlass clip for rapid windlass lock and bilateral buttress for added strength Enhanced windlass strap changed to tactical gray and secured to the latch for constant contact providing safe windlass security and a place to document the application time Reinforced stabilization plate with beveled contact bar Signature red elliptical tip (Red Tip Technology) provides visual cue during application Providing a distinct visual means to distinguish training devices from personal/operational life-saving devices