Learn More
Med Vet International McKesson Tourniquet Band, 18 Inch Length, 16-6334, 250/Box
Catalog No. 503045439
Encompass_Preferred
McKesson Tourniquet Strap 18 Inch Length Polyisoprene 18-inch x 1-inch Blue Polyisoprene Textured to help provide a slip-resistant surface Pre-cut and packaged for easy dispensing High elasticity and tear resistance Non-Sterile Single use
Catalog No. 50-304-5439 Supplier Med Vet International Supplier No. 464713BX
McKesson Tourniquet Strap 18 Inch Length Polyisoprene 18-inch x 1-inch Blue Polyisoprene Textured to help provide a slip-resistant surface Pre-cut and packaged for easy dispensing High elasticity and tear resistance Non-Sterile Single use