Learn More
Med Vet International C-A-T Combat Tourniquet 6.5 Inch Length Military Latex Free Nylon, 30-0001, Each
Catalog No. 503043309
Encompass_Preferred
Single routing buckle for faster application, decreased blood loss, effective slack removal, fewer windlass turns, and simplified training with single protocol application standards Reinforced windlass, stabilization plate, windlass clip and writeable windlass security strap Red Tip Technology elliptical tab provides added visual clues during application
Catalog No. 50-304-3309 Supplier Med Vet International Supplier No. 812819EA
Single routing buckle for faster application, decreased blood loss, effective slack removal, fewer windlass turns, and simplified training with single protocol application standards Reinforced windlass, stabilization plate, windlass clip and writeable windlass security strap Red Tip Technology elliptical tab provides added visual clues during application