Learn More
Med Vet International C-A-T Combat Tourniquet 6-1/2 Inch Length Nylon, 30-0023, Each
Catalog No. 503044124
Encompass_Preferred
Single Routing Buckle for faster application, decreased blood loss, effective slack removal, fewer windlass turns, and simplified training with single protocol application standards Reinforced windlass rod with increased diameter for enhanced strength and aggressive ribbing for improved grip Bilateral windlass clip for rapid windlass lock and bilateral buttress for added strength Enhanced windlass strap changed to tactical gray and secured to the latch for constant contact providing safe windlass security and a place to document the application time Reinforced stabilization plate with beveled contact bar Signature red elliptical tip (Red Tip Technology) provides visual cue during application Used by both conventional and Special Operation forces
Catalog No. 50-304-4124 Supplier Med Vet International Supplier No. 812821EA
Single Routing Buckle for faster application, decreased blood loss, effective slack removal, fewer windlass turns, and simplified training with single protocol application standards Reinforced windlass rod with increased diameter for enhanced strength and aggressive ribbing for improved grip Bilateral windlass clip for rapid windlass lock and bilateral buttress for added strength Enhanced windlass strap changed to tactical gray and secured to the latch for constant contact providing safe windlass security and a place to document the application time Reinforced stabilization plate with beveled contact bar Signature red elliptical tip (Red Tip Technology) provides visual cue during application Used by both conventional and Special Operation forces