Learn More
Med Vet International McKesson Tourniquet Band on Roll, 18 Inch Length, 16-6335, 500/Case
Catalog No. 503044403
Encompass_Preferred
McKesson Tourniquet Strap 18 Inch Length Rolled and Banded Perforated Roll 18-inch x 1-inch Blue Tetraphenylethylene (TPE) Perforated at 18-inch increments on roll for easy removal out of the dispenser box Textured to help provide a slip-resistant surface High elasticity and tear resistance Non-Sterile Single use
Catalog No. 50-304-4403 Supplier Med Vet International Supplier No. 520569CS
McKesson Tourniquet Strap 18 Inch Length Rolled and Banded Perforated Roll 18-inch x 1-inch Blue Tetraphenylethylene (TPE) Perforated at 18-inch increments on roll for easy removal out of the dispenser box Textured to help provide a slip-resistant surface High elasticity and tear resistance Non-Sterile Single use