Learn More
Medchemexpress LLC HY-109193 5mg , Tovinontrine CAS:2062661-53-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1091935MG
Encompass_Preferred
HY-109193 5mg Tovinontrine CAS: 2062661-53-2 Tovinontrine (IMR-687) is a highly potent and selective phosphodiesterase-9 (PDE9) inhibitor specifically for the treatment of sickle cell disease. IC50s are 8.19 nM and 9.99 nM for PDE9A1 and PDE9A2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1283