Learn More
Crescent Chemical Co Inc TOWBIN BUFFER 10X FOR WESTERN
SDP
Catalog No. 50248614
Encompass_Preferred
Towbin Buffer 10X for Western Blotting Supplied as 10 x concentrate (0.25 M Tris and 1.92 M glycine in aqueous solution).Working buffer: dilute 100 ml of 10x concentrate with 200 ml methanol and 700 ml distilled water. Size - 1L Storage Conditions - +15 °C
Catalog No. 50-248-614 Supplier Crescent Chemical Co Inc Supplier No. SE42558.02
Towbin Buffer 10X for Western Blotting Supplied as 10 x concentrate (0.25 M Tris and 1.92 M glycine in aqueous solution).Working buffer: dilute 100 ml of 10x concentrate with 200 ml methanol and 700 ml distilled water. Size - 1L Storage Conditions - +15 °C