Learn More
Med Vet International Miltex Vantage Backhaus Towel Clamp, 5.25in.
Catalog No. 502829864
Encompass_Preferred
The Miltex Vantage V97-506 Backhaus Towel Clamp is a perforating clamp used for grasping tissue, securing towels or drapes, and holding or reducing small bone fractures. This product is made of mid-grade surgical steel and measures 5.25in.
Catalog No. 50-282-9864 Supplier Med Vet International Supplier No. MILV97506
The Miltex Vantage V97-506 Backhaus Towel Clamp is a perforating clamp used for grasping tissue, securing towels or drapes, and holding or reducing small bone fractures. This product is made of mid-grade surgical steel and measures 5.25in.