Learn More
Med Vet International McKesson Sanitizing Skin Wipe, Unscented, 269, Individual Packet, Qty 1
Catalog No. 503045144
Encompass_Preferred
Sanitizing Skin Wipe McKesson Individual Packet BZK (Benzalkonium Chloride) Unscented. Designed for general antiseptic cleansing for both patients and clinicians Each towelette contains 0.13% w/v benzalkonium chloride to help disinfect Sterile towelettes are 5 x 7 inches
Catalog No. 50-304-5144 Supplier Med Vet International Supplier No. 865276EA
Sanitizing Skin Wipe McKesson Individual Packet BZK (Benzalkonium Chloride) Unscented. Designed for general antiseptic cleansing for both patients and clinicians Each towelette contains 0.13% w/v benzalkonium chloride to help disinfect Sterile towelettes are 5 x 7 inches