Learn More
Med Vet International Skin Barrier Wipe StingGone™ 20% Strength Polyaminopropyl Biguanide Individual Packet NonSterile, 1503, 50/Box
Catalog No. 503044981
Encompass_Preferred
Use to protect skin from adhesive stripping, body fluids, and incontinence Alcohol free Dimensions: 1-1/5 X 2-3/5 Inch Non-Sterile
Catalog No. 50-304-4981 Supplier Med Vet International Supplier No. 770594BX
Use to protect skin from adhesive stripping, body fluids, and incontinence Alcohol free Dimensions: 1-1/5 X 2-3/5 Inch Non-Sterile