Learn More
Brady™ Tower Skirt Lockable Confined Space Training Kits
Catalog No. 19129778 Shop All Brady Corporation Products
Change view
Click to view available options
Displayed Text:
Permit required. No unauthorized entry.
Dimensions (L x W):
24 x 24 in.
Catalog No. Displayed Text Dimensions (L x W)
19-129-778 Permit required. No unauthorized entry. 24 x 24 in.
1 options
Catalog No. 19-129-778 Supplier Brady™ Supplier No. 151088
Visually identifying your confined spaces will keep you compliant, but it may leave gaps in safety. Add another level of safety with this Confined Space Safety Cover to keep your crew safe.
  • Get the complete safety package – unlike signs and tapes this cover provides a sign and an unavoidable boundary in one easy-to-use product
  • Perfect for manways that need to be locked out or secured, but little or no flange is present
  • Lock it down – get another level of safety with lockable options that give authorized personnel the option to apply a padlock to maintain restrictive access
  • Includes (2) mesh covers and (2) ratchet straps
Specifications
Product Type Confined Space Cover
Color Black/Red
Material PVC
Dimensions (L x W) 24 x 24 in.
Displayed Text Permit required. No unauthorized entry.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.