Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Devices Molecular Devices Cell Integrity Kit, Fluorescent, Live/Dead, 96- and 384-Well

Catalog No. 5000014375
Encompass_Preferred

The Molecular Devices EarlyTox Cell Integrity Kit R8214 is a fluorescence-based assay for rapid differentiation of live and dead cells using complementary nuclear dyes. The bulk kit is formulated for high-throughput imaging in 96- and 384-well plates and supports a homogeneous mix-and-read workflow with an optional masking reagent for improved signal.

  • Sufficient for ten 96-well plates (approximately 960 wells) or equivalent 384-well plates.
  • Contains two nuclear dyes for live and dead cell discrimination and masking reagent for homogeneous assays.
  • Assay principle based on membrane permeability detection for live/dead identification.
  • Compatible with adherent and non-adherent cell types and high-throughput imaging workflows.
  • Recommended plate types: black-well, clear-bottom plates for imaging.

This item is not returnable. View return policy
Catalog No. 50-000-14375 Supplier Molecular Devices Supplier No. R8214
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Molecular Devices EarlyTox Cell Integrity Kit R8214 is a fluorescence-based assay for rapid differentiation of live and dead cells using complementary nuclear dyes. The bulk kit is formulated for high-throughput imaging in 96- and 384-well plates and supports a homogeneous mix-and-read workflow with an optional masking reagent for improved signal.

  • Sufficient for ten 96-well plates (approximately 960 wells) or equivalent 384-well plates.
  • Contains two nuclear dyes for live and dead cell discrimination and masking reagent for homogeneous assays.
  • Assay principle based on membrane permeability detection for live/dead identification.
  • Compatible with adherent and non-adherent cell types and high-throughput imaging workflows.
  • Recommended plate types: black-well, clear-bottom plates for imaging.

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.