Learn More
Quidel Triage TOX Drug Screen Panel
SDP
Catalog No. 1398229
Change view
Click to view available options
Quantity:
25 Tests/Pk.
Catalog No. Quantity
13-982-29 25 Tests/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 13-982-29 Supplier Quidel Triage Supplier No. 94600
Add to Cart
Add to Cart
This item is not returnable. View return policy
Quidel Triage TOX Drug Screen Panel is a fluorescence immunoassay to be used for the qualitative determination of the presence of drugs or major metabolites above the threshold concentrations of up to 9 distinct drug classes.
  • Reliable
  • Built-in QC satisfies daily QC, External POS/NEG QC run once per month
  • Accurate
  • Calibrated
  • Patient sample is automatically analyzed against the lot-specific
  • Multi-point calibration curve provided via the CODE CHIP™ module
  • Customizable panels with Test Select™
  • Qualitative POS/NEG results are displayed, printed and can be transmitted
  • Batch mode capability allows for increased throughput
Specifications
Content And Storage 2°C to 8°C
Detectable Analytes Amphetamine (AMP), Methamphetamine (mAMP), Barbiturates (BAR), Benzodiazepines (BZO), Cocaine (COC), Methadone (MTD), Opiates (OPI), Cannabinoids (THC), Tricyclic Antidepressants (TCA)
Quantity 25 Tests/Pk.