Learn More
Cpi International TOX PREPACKAGED COLUMN
SDP
Catalog No. NC3642000
Encompass
  • Designed for efficient total organic halogens (TOX) analysis.
  • Pre-filled with 40 mg of carbon for consistent performance.
  • 3 mm inner diameter for optimized flow rates.
  • Glass material ensures durability and chemical resistance.
  • Compatible with Thermo Euroglas and TE Instruments Xplorer analyzers.
  • Pack quantity of 100 for high-volume applications.
  • Features a white, 2-layer Cerafelt of 1/8 in thickness.
  • Length of 47 mm for compact integration into systems.
  • 6 mm outer diameter for standard fitting in various setups.
Catalog No. NC3642000 Supplier Cpi International Supplier No. 634004E
  • Designed for efficient total organic halogens (TOX) analysis.
  • Pre-filled with 40 mg of carbon for consistent performance.
  • 3 mm inner diameter for optimized flow rates.
  • Glass material ensures durability and chemical resistance.
  • Compatible with Thermo Euroglas and TE Instruments Xplorer analyzers.
  • Pack quantity of 100 for high-volume applications.
  • Features a white, 2-layer Cerafelt of 1/8 in thickness.
  • Length of 47 mm for compact integration into systems.
  • 6 mm outer diameter for standard fitting in various setups.