Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 27324MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: TOX3/1123. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b kappa. Positive Control: SH-SY5Y, MDA-MB-435 cells. Brain or breast or gastric Carcinoma. Cellular Staining: Cytoplasmic & Nuclear. Buffer: 1mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant fragment (139 Amino acid residues around aa 200-400) of human TOX3 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 63kDa.Chromosome: 16q12.1. Gene Symbol: TOX3. Gene ID: 27324. Ensemble: ENSG00000103460. Uniprot: NP_001073899.
Catalog No. 50-188-4747