Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toxaphene Solution, 2500μg/mL in Acetone, Ultra Scientific

Catalog No. USPPS2401
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-PPS-2401 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-PPS-2401 Supplier Agilent Technologies Supplier No. PPS2401
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5204-74-0 , 67-64-1
Molecular Formula C8H19NO2
Molecular Weight (g/mol) 161.245
InChI Key AVBGNFCMKJOFIN-UHFFFAOYSA-N
PubChem CID 165295
IUPAC Name acetic acid;N,N-diethylethanamine
SMILES CCN(CC)CC.CC(=O)O

Specifications

Physical Form Liquid
Color Colorless
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 2500μg/mL
Chemical Name or Material Toxaphene Solution, 2500μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.