Learn More
Toxic Substances Mix 1 (For EPA 625, CLP)
Catalog No. USUS103N
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
Catalog No. Quantity
US-US-103N 1 x 1 mL
1 options
Catalog No. US-US-103N Supplier Agilent Technologies Supplier No. US103N1
  • Page 307C, † Designates A2LA-Certified product.
  • Page 307Ca, † Designates A2LA-Certified product.
  • Page 307Cc, † Designates A2LA-Certified product.
Specifications
Chemical Name or Material Toxic Substances Mix 1
Packaging Ampule
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) Each component at 2000μg/mL in Methylene Chloride:Benzoic Acid2-Methylphenol4-Methylphenol2, 4, 5-Trichlorophenol