Learn More
Lonza Walkersville ToxiLight 100% Lysis Control Set, 200 Test
Supplier:  Lonza Walkersville LT07517
Encompass
The ToxiLight 100% Lysis Reagent set is intended for use with the ToxiLight Non-destructive Cytotoxicity Assay (sold seperately) and is designed to give the user a 100% lysis or total adenylate kinase control. It contains the 100% Lysis Reagent, 10 ml and Tris-Acetate Buffer, 50 ml (required volume correction for sample wells), and is suitable for 200 tests in a 96 well format.
Catalog No. NC9868118