Learn More
Cayman Chemical ToxIn A from ClostrIdIM dIF
SDP
Catalog No. 502982895
Encompass_Preferred
An enterotoxin that inactivates host Rho family GTPases by glucosylation, altering cytoskeletal structure, cell division, and cell-cell adhesion
Catalog No. 50-298-2895 Supplier Cayman Chemical Supplier No. 19664100 UG
Add to Cart
Add to Cart
An enterotoxin that inactivates host Rho family GTPases by glucosylation, altering cytoskeletal structure, cell division, and cell-cell adhesion