Learn More
Medchemexpress LLC Toxin B/TcdB C. dif 10ug | 10ug
SDP
Catalog No. 5000194673
Encompass_Preferred
Toxin B/TcdB protein is the precursor of the cytotoxin TcdB which can strategically destroy the colon epithelium and cause host inflammatory response and diarrhea in Clostridium difficile infection Compared with TcdA TcdB has higher virulence and is essential for eliciting host responses Toxin B/TcdB Protein C difficile (His) is the recombinant Toxin B/TcdB protein expressed by E coli with C-6 His labeled tag
Catalog No. 50-001-94673 Supplier Medchemexpress LLC Supplier No. HYP7921910UG
Toxin B/TcdB protein is the precursor of the cytotoxin TcdB which can strategically destroy the colon epithelium and cause host inflammatory response and diarrhea in Clostridium difficile infection Compared with TcdA TcdB has higher virulence and is essential for eliciting host responses Toxin B/TcdB Protein C difficile (His) is the recombinant Toxin B/TcdB protein expressed by E coli with C-6 His labeled tag