Learn More
Cayman Chemical ToxIn BClostrIdIM dIF 50ug
SDP
Catalog No. 502984245
Encompass_Preferred
A cytotoxin that inactivates host Rho family GTPases by glucosylation, altering cytoskeletal structure, cell division, and cell-cell adhesion
Catalog No. 50-298-4245 Supplier Cayman Chemical Supplier No. 1966550 UG
A cytotoxin that inactivates host Rho family GTPases by glucosylation, altering cytoskeletal structure, cell division, and cell-cell adhesion