Learn More
Genscript Corporation ToxinEraser Equilibration Buf
Catalog No. 5000058076
Encompass
b ToxinEraser sup TM /sup Equilibration Buffer /b (Cat No M01054) is suit for equilibration of ToxinEraser sup TM /sup Endotoxin Removal Kit and ToxinEraser sup TM /sup Endotoxin Removal Resin br li b Endotoxin level /b 0 015 EU/ml /li
Catalog No. 50-000-58076 Supplier Genscript Corporation Supplier No. M01054
b ToxinEraser sup TM /sup Equilibration Buffer /b (Cat No M01054) is suit for equilibration of ToxinEraser sup TM /sup Endotoxin Removal Kit and ToxinEraser sup TM /sup Endotoxin Removal Resin br li b Endotoxin level /b 0 015 EU/ml /li