Learn More
Genscript Corporation ToxinEraser Regeneration Buff
Catalog No. 5000057332
Encompass
b ToxinEraser sup TM /sup Regeneration Buffer /b (Cat No M01053) is suit for regeneration of ToxinEraser sup TM /sup Endotoxin Removal Kit and ToxinEraser sup TM /sup Endotoxin Removal Resin br b Endotoxin level /b 0 015 EU/ml
Catalog No. 50-000-57332 Supplier Genscript Corporation Supplier No. M01053
b ToxinEraser sup TM /sup Regeneration Buffer /b (Cat No M01053) is suit for regeneration of ToxinEraser sup TM /sup Endotoxin Removal Kit and ToxinEraser sup TM /sup Endotoxin Removal Resin br b Endotoxin level /b 0 015 EU/ml