Learn More
FUJIFILM Biosciences TOXINOMETER ET-7000/E
Catalog No. 5000450418
Encompass
16-well computer-operated kinetic incubating tube reader capable of conducting all three LAL methodologies
Catalog No. 50-004-50418 Supplier FUJIFILM Biosciences Supplier No. 29336061
16-well computer-operated kinetic incubating tube reader capable of conducting all three LAL methodologies